Merck
  • A Family of Vertebrate-Specific Polycombs Encoded by the LCOR/LCORL Genes Balance PRC2 Subtype Activities.
A Family of Vertebrate-Specific Polycombs Encoded by the LCOR/LCORL Genes Balance PRC2 Subtype Activities.
Molecular cell (2018-04-10)
Eric Conway, Emilia Jerman, Evan Healy, Shinsuke Ito, Daniel Holoch, Giorgio Oliviero, Orla Deevy, Eleanor Glancy, Darren J Fitzpatrick, Marlena Mucha, Ariane Watson, Alan M Rice, Paul Chammas, Christine Huang, Indigo Pratt-Kelly, Yoko Koseki, Manabu Nakayama, Tomoyuki Ishikura, Gundula Streubel, Kieran Wynne, Karsten Hokamp, Aoife McLysaght, Claudio Ciferri, Luciano Di Croce, Gerard Cagney, Raphaël Margueron, Haruhiko Koseki, Adrian P Bracken
ABSTRACT
The polycomb repressive complex 2 (PRC2) consists of core subunits SUZ12, EED, RBBP4/7, and EZH1/2 and is responsible for mono-, di-, and tri-methylation of lysine 27 on histone H3. Whereas two distinct forms exist, PRC2.1 (containing one polycomb-like protein) and PRC2.2 (containing AEBP2 and JARID2), little is known about their differential functions. Here, we report the discovery of a family of vertebrate-specific PRC2.1 proteins, "PRC2 associated LCOR isoform 1" (PALI1) and PALI2, encoded by the LCOR and LCORL gene loci, respectively. PALI1 promotes PRC2 methyltransferase activity in vitro and in vivo and is essential for mouse development. Pali1 and Aebp2 define mutually exclusive, antagonistic PRC2 subtypes that exhibit divergent H3K27-tri-methylation activities. The balance of these PRC2.1/PRC2.2 activities is required for the appropriate regulation of polycomb target genes during differentiation. PALI1/2 potentially link polycombs with transcriptional co-repressors in the regulation of cellular identity during development and in cancer.
Sigma-Aldrich
MISSION® TRC2 pLKO.5-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes
Sigma-Aldrich
Anti-DEDAF antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Trypsin Singles, Proteomics Grade
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Schneider′s Insect Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for insect cell culture
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)