Skip to Content
Merck
Warfarin Related Compound A certified reference material, pharmaceutical secondary standard

Warfarin Related Compound A

Warfarin Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
3-(2-Hydroxyphenyl)-5-phenyl-2-cyclohexen-1-one, Alice’s ketone
Empirical Formula (Hill Notation):
C18H16O2
Molecular Weight:
264.32

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade