Skip to Content
Merck
Trifluridine CRS European Pharmacopoeia (EP) Reference Standard

Trifluridine CRS

Trifluridine CRS European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Trifluorothymidine, α,α,α-Trifluorothymidine, 2′-Deoxy-5-trifluoromethyluridine, Trifluorothymine deoxyriboside, Trifluridine
Empirical Formula (Hill Notation):
C10H11F3N2O5
CAS Number:
Molecular Weight:
296.20

Products

Formula Weight
Manufacturer
Melting Point (°C)
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing