
TOYOPEARL® HP Size Exclusion Sampler, (HW-40, 50, 55, 65S), 30 μm particles

Synonym(s):
TOYOPEARL® Size Exclusion Sampler
Products
Manufacturer
Physical Form
Product Line
Separation Technique
Package Size
Particle Size (µm)
Technique
Showing 1-1 of 1