Skip to Content
Merck
TissueFab® photoinitiator LAP, low endotoxin

TissueFab® photoinitiator

TissueFab® photoinitiator LAP, low endotoxin
Synonym(s):
Lithium phenyl-2,4,6-trimethylbenzoylphosphinate
Empirical Formula (Hill Notation):
C16H16LiO3P
CAS Number:
Molecular Weight:
294.21

Products

Brand
Feature
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing