
Swainsonine

Synonym(s):
(1S,2R,8R,8aR)-1,2,8-Octahydroindolizidinetriol
Empirical Formula (Hill Notation):
C8H15NO3
CAS Number:
Molecular Weight:
173.21
Products
Brand
Biological Source
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing