Skip to Content
Merck
Sulpiride solution 1 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sulpiride solution

Sulpiride solution 1 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®
Synonym(s):
(±)-Sulpiride, (±)-5-(Aminosulfonyl)-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2-methoxybenzamide, (±)-N-1-(Ethylpyrrolidin-2-ylmethyl)-2-methoxy-5-sulfamoylbenzamide
Empirical Formula (Hill Notation):
C15H23N3O4S
CAS Number:
Molecular Weight:
341.43

Products

Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade