
Potassium hexachlororuthenate(IV)

Synonym(s):
Potassium hexachlororuthenate(IV)
Empirical Formula (Hill Notation):
K2RuCl6
CAS Number:
Molecular Weight:
391.98
Products
Brand
Color
Formula Weight
Physical Form
Quality Segment
Application
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing