Skip to Content
Merck
Potassium bis(trifluoromethylsulfonyl)imide ≥99.5% trace metals basis, anhydrous, battery grade

Potassium bis(trifluoromethylsulfonyl)imide

Potassium bis(trifluoromethylsulfonyl)imide ≥99.5% trace metals basis, anhydrous, battery grade
Synonym(s):
Potassium trifluoromethanesulfonimide, Potassium bis(trifluoromethanesulfonyl)imide
Empirical Formula (Hill Notation):
C2F6KNO4S2
CAS Number:
Molecular Weight:
319.24

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-1 of 1