
Penoxsulam Metabolite 5-OH

Synonym(s):
2-(2,2-Difluoroethoxy)-N-(5-hydroxy-8-methoxy-[1,2,4]triazolo[1,5-c]pyrimidin-2-yl)-6-(trifluoromethyl)benzenesulfonamide
Empirical Formula (Hill Notation):
C15H12F5N5O5S
Molecular Weight:
469.34
Products
Product Category
Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing