
Nequinate

Synonym(s):
Methyl 7-(benzyloxy)-6-butyl-1,4-dihydro-4-oxo-3-quinolinecarboxylate, Methyl benzoquate
Empirical Formula (Hill Notation):
C22H23NO4
Products
Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing