Skip to Content
Merck
vibrant-m

Lacosamide Related Compound F

vibrant-m
Synonym(s):
2-Acetamido-N-benzyl-3-hydroxypropionamide
Empirical Formula (Hill Notation):
C12H16N2O3
CAS Number:
Molecular Weight:
236.27

Products

Feature
Formula Weight
Agency
Quality Segment
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1