
Citalopram Related Compound D

Synonym(s):
Citalopram Related Compound D, 1-(4-Fluorophenyl)-1-(3-methylamino)propyl)-1,3-dihydroisobenzofuran-5-carbonitrile hydrochloride
Empirical Formula (Hill Notation):
C19H19FN2O · HCl
Molecular Weight:
346.83
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2