Skip to Content
Merck
vibrant-m

Calcium folinate for system suitiability A

vibrant-m
Synonym(s):
Folinic acid calcium salt, Calcium folinate
Empirical Formula (Hill Notation):
C20H21CaN7O7
CAS Number:
Molecular Weight:
511.50

Products

Formula Weight
Manufacturer
Application
Special Grade
Showing 1-1 of 1