Skip to Content
MY
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bulk Reagent - RHCE (Rh) Specificities
Products
Brand
Millipore (6)
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Quality Segment
Package Size
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
TW-1L-BK
Bulk Reagent - RHCE (Rh) Specificities,
BIOSCOT
™
Anti-C from the clone MS-273, Human IgM, Blood Grouping Reagent
Expand
View Pricing
TU-1L-BK
Bulk Reagent - RHCE (Rh) Specificities,
BIOSCOT
™
Anti-e from the clone MS-16/MS-21/MS-63, Human IgM, Blood Grouping Reagent
Expand
View Pricing
TA-1L-BK
Bulk Reagent - RHCE (Rh) Specificities,
BIOSCOT
™
Anti-E from the clone MS-80/MS-258, Human IgM, Blood Grouping Reagent
Expand
View Pricing
TX-1L-BK
Bulk Reagent - RHCE (Rh) Specificities,
BIOSCOT
™
Anti-C
w
from the clone MS-110, Human IgM, Blood Grouping Reagent
Expand
View Pricing
TK-1L-BK
Bulk Reagent - RHCE (Rh) Specificities,
BIOSCOT
™
Anti-c from the clone MS-33, Human IgM, Blood Grouping Reagent
Expand
View Pricing
TV-1L-BK
Bulk Reagent - RHCE (Rh) Specificities,
BIOSCOT
™
Anti-c from the clone MS-35, Human IgM, Blood Grouping Reagent
Expand
View Pricing