
β-Carboline-3-carboxylic acid N-methylamide

Synonym(s):
β-CCA methylamide, FG-7142, N-Methyl-β-carboline-3-carboxamide
Empirical Formula (Hill Notation):
C13H11N3O
CAS Number:
Molecular Weight:
225.25
Products
Product Category
Color
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing