Skip to Content
Merck
Azure II (C.I.52010/52015) for microscopy

Azure II (C.I.52010/52015)

Azure II (C.I.52010/52015) for microscopy
Synonym(s):
Azure II
Linear Formula:
C16H18N3S · C15H16N3S ·2 Cl
CAS Number:
Molecular Weight:
625.68

Products

Brand
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing