Skip to Content
MY
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-(Dimethylamino)ethyl methacrylate
Synonym(s):
Methacrylic acid 2-(dimethylamino)ethyl ester
Linear Formula:
CH
2
=C(CH
3
)COOCH
2
CH
2
N(CH
3
)
2
CAS Number:
2867-47-2
Molecular Weight:
157.21
EC Number:
220-688-8
Products
Product Category
acrylic monomers (1)
building blocks (1)
monomers (1)
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Monomer Type
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
234907
2-(Dimethylamino)ethyl methacrylate,
contains 700-1000 ppm monomethyl ether hydroquinone as inhibitor, 98%
Expand
View Pricing
Y0001489
2-(Dimethylamino)ethyl methacrylate,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing