Skip to Content
Merck
1,4-Dithiane-2,5-diol technical, ≥90% (S)

1,4-Dithiane-2,5-diol

1,4-Dithiane-2,5-diol technical, ≥90% (S)
Synonym(s):
2,5-Dihydroxy-1,4-dithiane, Mercaptoacetaldehyde dimer
Empirical Formula (Hill Notation):
C4H8O2S2
CAS Number:
Molecular Weight:
152.24

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing