Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
HOCH2CH2OH
CAS Number:
Molecular Weight:
62.07
Beilstein:
505945
EC Number:
MDL number:
UNSPSC Code:
12352137
PubChem Substance ID:
Grade:
reagent grade
Bp:
195-198 °C
Vapor pressure:
0.08 mmHg ( 20 °C)
grade
reagent grade
vapor density
2.1 (vs air)
vapor pressure
0.08 mmHg ( 20 °C)
product line
Vetec™
Assay
98%
form
liquid
autoignition temp.
752 °F
expl. lim.
15.3 %
dilution
(for analytical testing)
refractive index
n20/D 1.431 (lit.)
bp
195-198 °C
mp
−13 °C (lit.)
density
1.113 g/mL at 25 °C (lit.)
SMILES string
OCCO
InChI
1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2
InChI key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - STOT RE 2 Oral
Target Organs
Kidney
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
231.8 °F - closed cup
Flash Point(C)
111 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service