Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
Grade:
HPLC grade
Technique(s):
HPLC: suitable
Bp:
153 °C (lit.)
Vapor pressure:
2.7 mmHg ( 20 °C)
grade
HPLC grade
Quality Level
vapor density
2.5 (vs air)
vapor pressure
2.7 mmHg ( 20 °C)
product line
Vetec™
Assay
≥99.5%
form
liquid
autoignition temp.
833 °F
expl. lim.
15.2 %
technique(s)
HPLC: suitable
refractive index
n20/D 1.430 (lit.)
pH
6.7
bp
153 °C (lit.)
mp
−61 °C (lit.)
density
0.944 g/mL (lit.)
SMILES string
[H]C(=O)N(C)C
InChI
1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3
InChI key
ZMXDDKWLCZADIW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Solvent for many hydrophobic organic compounds.
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
135.5 °F - closed cup
Flash Point(C)
57.5 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service