Skip to Content
Merck
Sign Into View Organizational & Contract Pricing

About This Item

Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03
Beilstein:
3595639
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

LR
anhydrous

product line

Vetec

Assay

≥98%

form

solid

autoignition temp.

1112 °F

pH

7.5-9.0 (5% in water)

pKa 

4.76 (acetic acid)

mp

>300 °C (dec.) (lit.)

SMILES string

[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service