General description
SLK or LOSK is a Ste20-like protein kinase that associates with microtubule and centrosome. Inhibition of SLK activity by dominant-negative mutant or RNAi leads to unfocused microtubule arrangement indicating that SLK is needed for microtubule organization and for the proper localization of Golgi complex. Expression of v-Src can down-regulate SLK activity through the involvement of CK2 which can directly phosphorylate and inhibit SLK activity whereas inhibition of CK2 in v-Src-transformed cells results in normal SLK activity. CK2 and SLK can be co-localized in fibroblasts spreading on fibronectin-coated substrates.
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service