- Schizosaccharomyces pombe homologs of the Saccharomyces cerevisiae mitochondrial proteins Cbp6 and Mss51 function at a post-translational step of respiratory complex biogenesis.
Schizosaccharomyces pombe homologs of the Saccharomyces cerevisiae mitochondrial proteins Cbp6 and Mss51 function at a post-translational step of respiratory complex biogenesis.
Mitochondrion (2012-02-22)
Inge Kühl, Thomas D Fox, Nathalie Bonnefoy
PMID22349564
초록
Complexes III and IV of the mitochondrial respiratory chain contain a few key subunits encoded by the mitochondrial genome. In Saccharomyces cerevisiae, fifteen mRNA-specific translational activators control mitochondrial translation, of which five are conserved in Schizosaccharomyces pombe. These include homologs of Cbp3, Cbp6 and Mss51 that participate in translation and the post-translational steps leading to the assembly of respiratory complexes III and IV. In this study we show that in contrast to budding yeast, Cbp3, Cbp6 and Mss51 from S. pombe are not required for the translation of mitochondrial mRNAs, but fulfill post-translational functions, thus probably accounting for their conservation.