콘텐츠로 건너뛰기
Merck
  • Expression and significance of p53, rb, p21/waf-1, p16/ink-4a, and PTEN tumor suppressors in canine melanoma.
Expression and significance of p53, rb, p21/waf-1, p16/ink-4a, and PTEN tumor suppressors in canine melanoma.
Veterinary pathology (2002-07-20)
A Koenig, S R Bianco, S Fosmire, J Wojcieszyn, J F Modiano
초록
The role of tumor suppressor genes in the pathogenesis of canine melanoma is incompletely understood. The genes encoding the tumor suppressors p53, Rb, p21 (waf-1), p16 (ink-4a), and PTEN have been postulated to contribute to the pathogenesis of melanoma in humans and experimental animal models. To assess whether inactivation of these genes similarly contributes to the origin and progression of canine melanoma, we examined their expression in seven distinct canine melanoma cell lines and in 31 retrospective samples (representing 29 dogs) of spontaneous canine melanoma. Various patterns suggestive of loss of tumor suppressor function emerged in these cell lines. The most frequently observed abnormality was loss or significant reduction of p16 expression in six of seven cell lines and in 21 of 26 tumor samples. Loss or significant reduction of PTEN expression was seen in four of seven cell lines and in 13 of 27 tumor samples. Although p53 was detectable in all the cell lines and in 24 of 30 tumors, exclusion of p53 from the nuclear compartment was observed in each of the cell lines and in 18 of 25 tumor samples. These results indicate that loss of function of these tumor suppressor proteins is a common occurrence that may contribute to the origin of canine melanoma. In our sample population, abnormalities in the expression or localization of one or more tumor suppressor proteins occurred with similar frequency in malignant and benign tumors; thus, additional work is necessary to determine how these proteins may impact disease progression and response to therapy.