콘텐츠로 건너뛰기
Merck
  • Requirements for Jag1-Rbpj mediated Notch signaling during early mouse lens development.
Requirements for Jag1-Rbpj mediated Notch signaling during early mouse lens development.
Developmental dynamics : an official publication of the American Association of Anatomists (2012-01-26)
Tien T Le, Kevin W Conley, Timothy J Mead, Sheldon Rowan, Katherine E Yutzey, Nadean L Brown
초록
During vertebrate lens development, the lens placode in the embryonic ectoderm invaginates into a lens vesicle, which then separates from the surface epithelium, followed by two waves of fiber cell differentiation. In the mouse, multiple labs have shown that Jag1-Notch signaling is critically required during the second wave of lens fiber cell formation. However, Notch signaling appears to play no obvious role during lens induction or morphogenesis, although multiple pathway genes are expressed at these earlier stages. Here, we explored functions for Notch signaling specifically during early lens development, by using the early-acting AP2α-Cre driver to delete Jag1 or Rbpj. We found that Jag1 and Rbpj are not required during lens induction, but are necessary for proper lens vesicle separation from the surface ectoderm. We conclude that precise levels of Notch signaling are essential during lens vesicle morphogenesis. In addition, AP2α-Cre-mediated deletion of Rbpj resulted in embryos with cardiac outflow tract and liver deformities, and perinatal lethality.