콘텐츠로 건너뛰기
Merck
  • Semaphorin 4D/Plexin-B1-mediated R-Ras GAP activity inhibits cell migration by regulating beta(1) integrin activity.
Semaphorin 4D/Plexin-B1-mediated R-Ras GAP activity inhibits cell migration by regulating beta(1) integrin activity.
The Journal of cell biology (2006-05-17)
Izumi Oinuma, Hironori Katoh, Manabu Negishi
초록
Plexins are cell surface receptors for semaphorins and regulate cell migration in many cell types. We recently reported that the semaphorin 4D (Sema4D) receptor Plexin-B1 functions as a GTPase-activating protein (GAP) for R-Ras, a member of Ras family GTPases implicated in regulation of integrin activity and cell migration. We characterized the role of R-Ras downstream of Sema4D/Plexin-B1 in cell migration. Activation of Plexin-B1 by Sema4D suppressed the ECM-dependent R-Ras activation, R-Ras-mediated phosphatydylinositol 3-kinase activation, and beta(1) integrin activation through its R-Ras GAP domain, leading to inhibition of cell migration. In addition, inactivation of R-Ras by overexpression of the R-Ras-specific GAP or knockdown of R-Ras by RNA interference was sufficient for suppressing beta(1) integrin activation and cell migration in response to the ECM stimulation. Thus, we conclude that R-Ras activity is critical for ECM-mediated beta(1) integrin activation and cell migration and that inactivation of R-Ras by Sema4D/Plexin-B1-mediated R-Ras GAP activity controls cell migration by modulating the activity of beta(1) integrins.