콘텐츠로 건너뛰기
Merck
  • Sphingosine 1-phosphate regulates IL-8 expression and secretion via S1PR1 and S1PR2 receptors-mediated signaling in extravillous trophoblast derived HTR-8/SVneo cells.
Sphingosine 1-phosphate regulates IL-8 expression and secretion via S1PR1 and S1PR2 receptors-mediated signaling in extravillous trophoblast derived HTR-8/SVneo cells.
Placenta (2015-09-01)
Daniela Brünnert, Svea Piccenini, Jens Ehrhardt, Marek Zygmunt, Pankaj Goyal
초록
Both villous and extravillous trophoblast (EVT) cells produce a wide range of cytokines and also respond to them in autocrine and paracrine manner. Deregulation of cytokine secretion may lead to various pathologic conditions including preeclampsia. IL-8, a pro-inflammatory cytokine, regulates various cellular functions such as neutrophil trafficking, cell adhesion, tumor growth and has a role in placental development. IL-8 also promotes trophoblast cell migration and invasion, and stimulates the secretion of progesterone. The induction and mechanism of IL-8 secretion by EVT is still unknown. IL-8 mRNA expression and secretion was determined using real-time PCR and ELISA respectively. To identify the mechanism of IL-8 expression and secretion, selective antagonists and agonist of S1P receptor subtypes, Rac1 and Rho-kinase inhibitors were used. We found that S1P induces IL-8 gene expression and protein secretion in EVT derived HTR-8/SVneo cells but not in BeWo cells. SEW2781, the selective agonist of S1PR(1), induced IL-8 gene expression but not protein secretion. The specific S1PR(2) inhibitor JTE-013 could drastically inhibit IL-8 secretion. Furthermore, pre-treatment of cells with the selective S1PR(1)/S1PR(3) antagonist VPC23019 inhibited IL-8 secretion by ∼45%. Selective Rho-kinase inhibitor Y27632 and Rac1 inhibitor NSC23766 could block IL-8 secretion in these cells. In this study, we could show for the first time that S1P induces IL-8 mRNA expression and protein secretion in EVT cell line. S1P-induced IL-8 gene expression is mainly regulated via S1PR(1) and its secretion is regulated through S1PR(2) receptor subtype. Rho GTPases signaling is essential for S1P-induced IL-8 secretion.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
SEW2871, ≥98% (HPLC), solid