콘텐츠로 건너뛰기
Merck
  • Distinction between MOG antibody-positive and AQP4 antibody-positive NMO spectrum disorders.
Distinction between MOG antibody-positive and AQP4 antibody-positive NMO spectrum disorders.
Neurology (2014-01-15)
Douglas Kazutoshi Sato, Dagoberto Callegaro, Marco Aurelio Lana-Peixoto, Patrick J Waters, Frederico M de Haidar Jorge, Toshiyuki Takahashi, Ichiro Nakashima, Samira Luisa Apostolos-Pereira, Natalia Talim, Renata Faria Simm, Angelina Maria Martins Lino, Tatsuro Misu, Maria Isabel Leite, Masashi Aoki, Kazuo Fujihara
초록
To evaluate clinical features among patients with neuromyelitis optica spectrum disorders (NMOSD) who have myelin oligodendrocyte glycoprotein (MOG) antibodies, aquaporin-4 (AQP4) antibodies, or seronegativity for both antibodies. Sera from patients diagnosed with NMOSD in 1 of 3 centers (2 sites in Brazil and 1 site in Japan) were tested for MOG and AQP4 antibodies using cell-based assays with live transfected cells. Among the 215 patients with NMOSD, 7.4% (16/215) were positive for MOG antibodies and 64.7% (139/215) were positive for AQP4 antibodies. No patients were positive for both antibodies. Patients with MOG antibodies represented 21.1% (16/76) of the patients negative for AQP4 antibodies. Compared with patients with AQP4 antibodies or patients who were seronegative, patients with MOG antibodies were more frequently male, had a more restricted phenotype (optic nerve more than spinal cord), more frequently had bilateral simultaneous optic neuritis, more often had a single attack, had spinal cord lesions distributed in the lower portion of the spinal cord, and usually demonstrated better functional recovery after an attack. Patients with NMOSD with MOG antibodies have distinct clinical features, fewer attacks, and better recovery than patients with AQP4 antibodies or patients seronegative for both antibodies.