콘텐츠로 건너뛰기
Merck
  • FoxP3 and indoleamine 2,3-dioxygenase immunoreactivity in sentinel nodes from melanoma patients.
FoxP3 and indoleamine 2,3-dioxygenase immunoreactivity in sentinel nodes from melanoma patients.
American journal of otolaryngology (2014-09-13)
Marisa Ryan, Jennifer Crow, Russel Kahmke, Samuel R Fisher, Zuowei Su, Walter T Lee
초록
1) Assess FoxP3/indoleamine 2,3-dioxygenase immunoreactivity in head and neck melanoma sentinel lymph nodes and 2) correlate FoxP3/indoleamine 2,3-dioxygenase with sentinel lymph node metastasis and clinical recurrence. Retrospective cohort study. Patients with sentinel lymph node biopsy for head and neck melanoma between 2004 and 2011 were identified. FoxP3/indoleamine 2,3-dioxygenase prevalence and intensity were determined from the nodes. Poor outcome was defined as local, regional or distant recurrence. The overall immunoreactivity score was correlated with clinical recurrence and sentinel lymph node metastasis using the chi-square test for trend. Fifty-six sentinel lymph nodes were reviewed, with 47 negative and 9 positive for melanoma. Patients with poor outcomes had a statistically significant trend for higher immunoreactivity scores (p=0.03). Positive nodes compared to negative nodes also had a statistically significant trend for higher immunoreactivity scores (p=0.03). Among the negative nodes, there was a statistically significant trend for a poor outcome with higher immunoreactivity scores (p=0.02). FoxP3/indoleamine 2,3-dioxygenase immunoreactivity correlates with sentinel lymph node positivity and poor outcome. Even in negative nodes, higher immunoreactivity correlated with poor outcome. Therefore higher immunoreactivity may portend a worse prognosis even without metastasis in the sentinel lymph node. This could identify a subset of patients that may benefit from future trials and treatment for melanoma through Treg and IDO suppression.