콘텐츠로 건너뛰기
Merck
  • Protective effect of madecassoside against cognitive impairment induced by D-galactose in mice.
Protective effect of madecassoside against cognitive impairment induced by D-galactose in mice.
Pharmacology, biochemistry, and behavior (2014-08-12)
Xing Lin, Shijun Zhang, Renbin Huang, Ling Wei, Shimei Tan, Chunhong Liang, Shujuan Lv, Yongxin Chen, Shuang Liang, Yuanchun Tian, Zhongpeng Lu, Quanfang Huang
초록
This study was designed to investigate the protective effect of madecassoside from Hydrocotyle sibthorpioides against cognitive impairment induced by D-galactose (D-gal) in mice. The result revealed that treatment with madecassoside significantly reversed D-gal-induced learning and memory impairments, as measured by the Morris water-maze test. Studies on the potential mechanisms of this action showed that madecassoside significantly reduced oxidative stress and suppress inflammatory responses via blocking NF-κB and ERK/p38 MAPK pathways. Moreover, madecassoside markedly attenuated the content and deposition of β-amyloid peptide by inducing a decrease in the expression of amyloid protein precursor, β-site amyloid cleaving enzyme-1 and cathepsin B and an increase in the levels of neprilysin and insulin-degrading enzyme. Madecassoside significantly increased the expression of synapse plasticity-related proteins in the hippocampus, such as postsynaptic density 95, long-term potentiation, N-methyl-D-aspartic acid receptors, Ca(2+)/calmodulin-dependent protein kinase II, NMDA receptor subunit 1, protein kinase C, protein kinase A, cAMP-response element binding protein, and brain-derived neurotrophic factor. In addition, madecassoside significantly increased the levels of acetylcholine but decreased cholinesterase activity. In conclusion, the protective effect of madecassoside against d-gal-induced cognitive impairment was mainly due to its ability to reduce oxidative damage, improve synaptic plasticity and restore cholinergic function. These findings suggest that madecassoside can be considered as a potential agent for preventing cognitive impairment.
MATERIALS
제품 번호
브랜드
제품 설명
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%