콘텐츠로 건너뛰기
Merck
  • Effects of electroacupuncture on ethanol-induced impairments of spatial learning and memory and Fos expression in the hippocampus in rats.
Effects of electroacupuncture on ethanol-induced impairments of spatial learning and memory and Fos expression in the hippocampus in rats.
Neuroscience letters (2014-06-14)
Bin Lu, Zhao Ma, Fei Cheng, Yan Zhao, Xin Zhang, Huijuan Mao, Xueyong Shen, Sheng Liu
초록
It is well established that alcohol impairs spatial learning and memory. Here, we investigated the effects of electroacupuncture (EA) at ST36 or nonacupoint on ethanol-induced learning and memory impairment and the expression of Fos in the hippocampus. Ethanol (5g/kg) was administered intragastrically once a day for 5 consecutive days; 2Hz EA was administered immediately after ethanol exposure. After a 2-day ethanol abstinence, for 6 consecutive days, the rats were submitted to Morris water maze training. Probe trials were performed on 1 day after the final training session. We also applied immunohistochemistry to detect Fos-positive nuclei in the hippocampus. We found that 5-day ethanol exposure markedly decreased spatial learning and memory abilities in the Morris water maze task as indicated by escape latency and time in the target quadrant. EA treatment shortened the time of reaching platform and increased times traveled in the target quadrant (P<0.05). Animals administered with ethanol emitted significantly fewer Fos expression in the hippocampal CA1 area. EA increased Fos expression in the hippocampal CA1 area. Significant correlations were obtained between Fos protein expression in CA1 and time in the target quadrant. Altogether, these results suggest that EA protects against ethanol-induced impairments of spatial learning and memory, which may be involved in the hippocampal CA1 area. EA treatment may provide a novel nonpharmacological strategy for ethanol-induced learning and memory impairment.