콘텐츠로 건너뛰기
Merck
  • CTRP5 ameliorates palmitate-induced apoptosis and insulin resistance through activation of AMPK and fatty acid oxidation.
CTRP5 ameliorates palmitate-induced apoptosis and insulin resistance through activation of AMPK and fatty acid oxidation.
Biochemical and biophysical research communications (2014-09-10)
Won-Mo Yang, Wan Lee
초록
Lipotoxicity resulting from a high concentration of saturated fatty acids is closely linked to development of insulin resistance, as well as apoptosis in skeletal muscle. CTRP5, an adiponectin paralog, is known to activate AMPK and fatty acid oxidation; however, the effects of CTRP5 on palmitate-induced lipotoxicity in myocytes have not been investigated. We found that globular domain of CTRP5 (gCTRP5) prevented palmitate-induced apoptosis and insulin resistance in myocytes by inhibiting the activation of caspase-3, reactive oxygen species accumulation, and IRS-1 reduction. These beneficial effects of gCTRP5 are mainly attributed to an increase in fatty acid oxidation through phosphorylation of AMPK. These results provide a novel function of CTRP5, which may have preventive and therapeutic potential in management of obesity, insulin resistance, and type 2 diabetes mellitus.