콘텐츠로 건너뛰기
Merck
  • Induction of inflammation by West Nile virus capsid through the caspase-9 apoptotic pathway.
Induction of inflammation by West Nile virus capsid through the caspase-9 apoptotic pathway.
Emerging infectious diseases (2002-12-25)
Joo-Sung Yang, Mathura P Ramanathan, Karuppiah Muthumani, Andrew Y Choo, Sung-Ha Jin, Qian-Chun Yu, Daniel S Hwang, Daniel K Choo, Mark D Lee, Kesen Dang, Waixing Tang, J Joseph Kim, David B Weiner
초록
West Nile virus (WNV) is a member of the Flaviviridae family of vector-borne pathogens. Clinical signs of WNV infection include neurologic symptoms, limb weakness, and encephalitis, which can result in paralysis or death. We report that the WNV-capsid by itself induces rapid nuclear condensation and cell death in tissue culture. Apoptosis is induced through the mitochondrial pathway resulting in caspase-9 activation and downstream caspase-3 activation. Capsid gene delivery into the striatum of mouse brain or interskeletal muscle resulted in cell death and inflammation, likely through capsid-induced apoptosis in vivo. These studies demonstrate that the capsid protein of WNV may be responsible for aspects of viral pathogenesis through induction of the apoptotic cascade.