콘텐츠로 건너뛰기
Merck
  • A mutation in the human norepinephrine transporter gene (SLC6A2) associated with orthostatic intolerance disrupts surface expression of mutant and wild-type transporters.
A mutation in the human norepinephrine transporter gene (SLC6A2) associated with orthostatic intolerance disrupts surface expression of mutant and wild-type transporters.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2003-06-14)
Maureen K Hahn, David Robertson, Randy D Blakely
초록
The norepinephrine transporter (NET) mediates reuptake of norepinephrine released from neurons, and, as such, it is an important regulator of noradrenergic neurotransmission. Recently, our laboratory reported a polymorphism in the human NET (hNET) gene A457P in an individual with the autonomic disorder orthostatic intolerance (OI). The presence of the hNET-A457P allele tracked with elevated heart rates and plasma NE levels in family members. hNET-A457P lacks >98% transport activity in several heterologous expression systems. In the present work, Western blot and biotinylation analyses performed in transiently transfected COS-7 cells revealed impairment in processing of hNET-A457P to the fully glycosylated form and a decrease in surface expression to approximately 30% of hNET-wild type (hNET-wt). Because the hNET-A457P mutation is carried on a single allele in OI subjects, we examined the influence of cotransfection of hNET-wt and hNET-A457P and found that hNET-A457P exerts a dominant-negative effect on hNET-wt uptake activity. Experiments to determine oligomerization as a potential mechanism of the dominant-negative effect demonstrated that hNET-A457P coimmunoprecipitates with, and diminishes surface expression of, hNET-wt. These results reveal that hNET-A457P causes a conformational disruption that interferes with transporter biosynthetic progression and trafficking of both the mutant transporter and hNET-wt. These results elucidate a molecular mechanism for the disrupted NE homeostasis and cardiovascular function evident in OI patients with the hNET-A457P mutation.