뒤로
Valco
®
Precision Sampling Syringe, Series A-2, Removable needle
Products
배송
오늘 출고 (1)
제품 카테고리
syringes (9)
Manufacturer
길이(in)
용적(ml)
Volume (µL)
니들 모양
포장 단위
제품 필터
Showing 1-9 of 9
비교
제품 번호
설명
가격
22268
Valco
®
Precision Sampling Syringe, Series A-2, Removable needle,
volume 25 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.028 in. × 0.005 in.
확장
가격 보기
22273
Valco
®
Precision Sampling Syringe, Series A-2, Removable needle,
volume 1000 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in.
확장
가격 보기
22272
Valco
®
Precision Sampling Syringe, Series A-2, Removable needle,
volume 500 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in.
확장
가격 보기
22270-U
Valco
®
Precision Sampling Syringe, Series A-2, Removable needle,
volume 100 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.028 in. × 0.005 in.
확장
가격 보기
22271
Valco
®
Precision Sampling Syringe, Series A-2, Removable needle,
volume 250 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in.
확장
가격 보기
22275
Valco
®
Precision Sampling Syringe, Series A-2, Removable needle,
volume 5 mL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in.
확장
가격 보기
22274
Valco
®
Precision Sampling Syringe, Series A-2, Removable needle,
volume 2000 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in.
확장
가격 보기
22276
Valco
®
Precision Sampling Syringe, Series A-2, Removable needle,
volume 10 mL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in.
확장
가격 보기
22269-U
Valco
®
Precision Sampling Syringe, Series A-2, Removable needle,
volume 50 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.028 in. × 0.005 in.