뒤로
Hellma
®
absorption cuvettes, cells for magnetic stirrers
Products
제품 카테고리
cuvettes (2)
Manufacturer
직경(mm)
높이(mm)
Volume (µL)
검출 방법
Technique
제품 필터
Showing 1-2 of 2
비교
제품 번호
설명
가격
Z803669
Hellma
®
absorption cuvettes, cells for magnetic stirrers,
High Performance Quartz Glass, spectral range 200-2500 nm, pathlength 10 mm, chamber volume 3500 μL
확장
가격 보기
Z803774
Hellma
®
absorption cuvettes, cells for magnetic stirrers,
High Performance Quartz Glass, spectral range 200-2500 nm, pathlength 10 mm, chamber volume 1500 μL