Merck
  • A New Pentiptycene-Based Dianhydride and Its High-Free-Volume Polymer for Carbon Dioxide Removal.
A New Pentiptycene-Based Dianhydride and Its High-Free-Volume Polymer for Carbon Dioxide Removal.
ChemSusChem (2017-11-07)
Ahmad Arabi Shamsabadi, Farzad Seidi, Mohammad Nozari, Masoud Soroush
ABSTRACT
In addition to possessing excellent chemical, mechanical, and thermal stability, polyimides and polyetherimides have excellent solubility in many solvents, which renders them suitable for membrane preparation. Two new monomers [a pentiptycene-based dianhydride (PPDAn) and a pentiptycene imide-containing diamine (PPImDA)] and a pentiptycene-based polyimide [PPImDA-4,4'-hexafluoroisopropylidene diphthalic anhydride (PPImDA-6FDA)] have been synthesized and characterized by FTIR and
Sigma-Aldrich
4-Fluorophthalic anhydride, 97%
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
1,2,4,5-Tetramethylbenzene, 98%
Sigma-Aldrich
p-Benzoquinone, reagent grade, ≥98%
Sigma-Aldrich
Sulfolane, 99%
Sigma-Aldrich
Sodium hydrosulfite, technical grade