Merck
  • Advanced Clinical Imaging and Tissue-based Biomarkers of the Eye for Toxicology Studies in Minipigs.
Advanced Clinical Imaging and Tissue-based Biomarkers of the Eye for Toxicology Studies in Minipigs.
Toxicologic pathology (2015-12-19)
Elke-Astrid Atzpodien, Bjoern Jacobsen, Juergen Funk, Bernd Altmann, Manuel A Silva Munoz, Thomas Singer, Cyrill Gyger, Pascal Hasler, Peter Maloca
ABSTRACT
There is increased interest to use minipigs in ocular toxicology studies due to their anatomical similarities with human eyes and as a substitute for nonhuman primates. This requires adaptation of enhanced optical coherence tomography (OCT) techniques and of ocular relevant immunohistochemistry (IHC) or in situ hybridization (ISH) markers to porcine eyes. In this study, OCT and OCT angiography (AngioOCT) were performed on adult Göttingen minipigs. To increase structural information on retinal and choroidal vasculature, OCT data were speckle denoized and choroidal blood vessels were segmented with threshold filtering. In addition, we established a set of IHC and ISH markers on Davidson's fixed paraffin-embedded minipig eyes: neurofilament-160, neuronal nuclei, calretinin, protein kinase C-α, vimentin, glial fibrillary acidic protein, glutamine synthetase, ionized calcium-binding adaptor molecule-1, rhodopsin, synaptophysin, postsynaptic density protein-95, retinal pigment epithelium (RPE)-specific protein-65, von Willebrand factor, α-smooth muscle actin, desmin, and Ki-67, thus enabling visualization of retinal neuronal and glial cells, photoreceptors, synapses, RPE, blood vessels, myocytes, macrophages, or cell proliferation. Using ISH, transcripts of vascular endothelial growth factor A, angiopoietin-2, and endothelial tyrosine kinase were visualized. This article describes for the first time in minipig eyes speckle noise-free OCT, AngioOCT, and a set of IHC/ISH markers on Davidson's fixed paraffin-embedded tissues and helps to establish the minipig for ocular toxicology and pharmacology studies.
Sigma-Aldrich
Anti-Retinal Pigment Epithelium 65 Antibody, clone 401.8B11.3D9, Chemicon®, from mouse
Sigma-Aldrich
Anti-Rhodopsin Antibody, clone RET-P1, clone RET-P1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein antibody, Mouse monoclonal, clone G-A-5, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Neurofilament 160 kDa Antibody, clone NN18, clone NN18, Chemicon®, from mouse
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glutamine Synthetase Antibody, clone GS-6, clone GS-6, Chemicon®, from mouse