Merck
  • Intrinsic antiproliferative activity of the innate sensor STING in T lymphocytes.
Intrinsic antiproliferative activity of the innate sensor STING in T lymphocytes.
The Journal of experimental medicine (2017-05-10)
Silvia Cerboni, Nadia Jeremiah, Matteo Gentili, Ulf Gehrmann, Cécile Conrad, Marie-Claude Stolzenberg, Capucine Picard, Bénédicte Neven, Alain Fischer, Sébastian Amigorena, Frédéric Rieux-Laucat, Nicolas Manel
ABSTRACT
Activation of the cyclic dinucleotide sensor stimulator of interferon (IFN) genes (STING) is critical for IFN and inflammatory gene expression during innate immune responses. However, the role of STING in adaptive immunity is still unknown. In this study, we show that STING activation reduces the proliferation of T lymphocytes. This activity was independent of TBK1 and IRF3 recruitment and of type I IFN but required a distinct C-terminal domain of STING that activates NF-κB. Inhibition of cell proliferation by STING required its relocalization to the Golgi apparatus and caused mitotic errors. T lymphocytes from patients carrying constitutive active mutations in TMEM173 encoding STING showed impaired proliferation and reduced numbers of memory cells. Endogenous STING inhibited proliferation of mouse T lymphocytes. Therefore, STING, a critical innate sensor, also functions intrinsically in cells of the adaptive immune system to inhibit proliferation.
Sigma-Aldrich
DMXAA, ≥98% (HPLC), solid
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
Lectin from Phaseolus vulgaris, lyophilized, powder
Sigma-Aldrich
Anti-MB21D1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Interferon-α/β Receptor Chain 2 Antibody, clone MMHAR-2, clone MMHAR-2, Chemicon®, from mouse
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution