Merck
The multifaceted roles of Leptospira enolase.
Research in microbiology (2016-12-19)
Natália Salazar, Matilde Costa Lima de Souza, Amanda Gameiro Biasioli, Ludmila Bezerra da Silva, Angela Silva Barbosa
ABSTRACT
A previous study had demonstrated that Leptospira enolase is secreted extracellularly by a yet unknown mechanism and reassociates with the bacterial membrane. Surface-anchored leptospiral enolase displays plasminogen binding activity. In this work, we explored the consequences of this interaction and also assessed whether Leptospira enolase might display additional moonlighting functions by interacting with other host effector proteins. We first demonstrated that enolase-bound plasminogen is converted to its active form, plasmin. The protease plasmin targets human fibrinogen and vitronectin, but not the complement proteins C3b and C5. Leptospira enolase also acts as an immune evasion protein by interacting with the negative complement regulators C4b binding protein and factor H. Once bound to enolase, both regulators remain functional as cofactors of factor I, mediating cleavage of C4b and C3b. In conclusion, enolase may facilitate leptospiral survival and dissemination, thus contributing to bacterial virulence. The identification and characterization of moonlighting proteins is a growing field of bacterial pathogenesis, as these multifaceted proteins may represent potential future therapeutic targets to fight bacterial infections.
Sigma-Aldrich
Vitronectin from human plasma, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Urokinase from human kidney cells, lyophilized powder
Sigma-Aldrich
Fibrinogen from human plasma, 50-70% protein (≥80% of protein is clottable)
Sigma-Aldrich
D-Val-Leu-Lys p-nitroanilide dihydrochloride, plasmin substrate
Sigma-Aldrich
Plasminogen activator inhibitor 1 (PAI-1) human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)