Merck
  • POPDC1(S201F) causes muscular dystrophy and arrhythmia by affecting protein trafficking.
POPDC1(S201F) causes muscular dystrophy and arrhythmia by affecting protein trafficking.
The Journal of clinical investigation (2015-12-08)
Roland F R Schindler, Chiara Scotton, Jianguo Zhang, Chiara Passarelli, Beatriz Ortiz-Bonnin, Subreena Simrick, Thorsten Schwerte, Kar-Lai Poon, Mingyan Fang, Susanne Rinné, Alexander Froese, Viacheslav O Nikolaev, Christiane Grunert, Thomas Müller, Giorgio Tasca, Padmini Sarathchandra, Fabrizio Drago, Bruno Dallapiccola, Claudio Rapezzi, Eloisa Arbustini, Francesca Romana Di Raimo, Marcella Neri, Rita Selvatici, Francesca Gualandi, Fabiana Fattori, Antonello Pietrangelo, Wenyan Li, Hui Jiang, Xun Xu, Enrico Bertini, Niels Decher, Jun Wang, Thomas Brand, Alessandra Ferlini
ABSTRACT
The Popeye domain-containing 1 (POPDC1) gene encodes a plasma membrane-localized cAMP-binding protein that is abundantly expressed in striated muscle. In animal models, POPDC1 is an essential regulator of structure and function of cardiac and skeletal muscle; however, POPDC1 mutations have not been associated with human cardiac and muscular diseases. Here, we have described a homozygous missense variant (c.602C>T, p.S201F) in POPDC1, identified by whole-exome sequencing, in a family of 4 with cardiac arrhythmia and limb-girdle muscular dystrophy (LGMD). This allele was absent in known databases and segregated with the pathological phenotype in this family. We did not find the allele in a further screen of 104 patients with a similar phenotype, suggesting this mutation to be family specific. Compared with WT protein, POPDC1(S201F) displayed a 50% reduction in cAMP affinity, and in skeletal muscle from patients, both POPDC1(S201F) and WT POPDC2 displayed impaired membrane trafficking. Forced expression of POPDC1(S201F) in a murine cardiac muscle cell line (HL-1) increased hyperpolarization and upstroke velocity of the action potential. In zebrafish, expression of the homologous mutation (popdc1(S191F)) caused heart and skeletal muscle phenotypes that resembled those observed in patients. Our study therefore identifies POPDC1 as a disease gene causing a very rare autosomal recessive cardiac arrhythmia and LGMD, expanding the genetic causes of this heterogeneous group of inherited rare diseases.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-POPDC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Actin Cytoskeleton / Focal Adhesion Staining Kit, The Actin Cytoskeleton & Focal Adhesion Staining Kit consists of TRITC-conjugated phalloidin, anti-Vinculin & DAPI for the immunofluorescent staining of actin filaments in the cytoskeleton as well as the nucleus of the cells.
Sigma-Aldrich
Monoclonal Anti-Dystrophin antibody produced in mouse, clone MANDRA1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid