Merck
  • Divergent Phenotypes of Human Regulatory T Cells Expressing the Receptors TIGIT and CD226.
Divergent Phenotypes of Human Regulatory T Cells Expressing the Receptors TIGIT and CD226.
Journal of immunology (Baltimore, Md. : 1950) (2015-05-23)
Christopher A Fuhrman, Wen-I Yeh, Howard R Seay, Priya Saikumar Lakshmi, Gaurav Chopra, Lin Zhang, Daniel J Perry, Stephanie A McClymont, Mahesh Yadav, Maria-Cecilia Lopez, Henry V Baker, Ying Zhang, Yizheng Li, Maryann Whitley, David von Schack, Mark A Atkinson, Jeffrey A Bluestone, Todd M Brusko
ABSTRACT
Regulatory T cells (Tregs) play a central role in counteracting inflammation and autoimmunity. A more complete understanding of cellular heterogeneity and the potential for lineage plasticity in human Treg subsets may identify markers of disease pathogenesis and facilitate the development of optimized cellular therapeutics. To better elucidate human Treg subsets, we conducted direct transcriptional profiling of CD4(+)FOXP3(+)Helios(+) thymic-derived Tregs and CD4(+)FOXP3(+)Helios(-) T cells, followed by comparison with CD4(+)FOXP3(-)Helios(-) T conventional cells. These analyses revealed that the coinhibitory receptor T cell Ig and ITIM domain (TIGIT) was highly expressed on thymic-derived Tregs. TIGIT and the costimulatory factor CD226 bind the common ligand CD155. Thus, we analyzed the cellular distribution and suppressive activity of isolated subsets of CD4(+)CD25(+)CD127(lo/-) T cells expressing CD226 and/or TIGIT. We observed TIGIT is highly expressed and upregulated on Tregs after activation and in vitro expansion, and is associated with lineage stability and suppressive capacity. Conversely, the CD226(+)TIGIT(-) population was associated with reduced Treg purity and suppressive capacity after expansion, along with a marked increase in IL-10 and effector cytokine production. These studies provide additional markers to delineate functionally distinct Treg subsets that may help direct cellular therapies and provide important phenotypic markers for assessing the role of Tregs in health and disease.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Millipore
MILLIPLEX® Human TH17 Magnetic Bead Panel Premixed - 25 Plex - Immunology Multiplex Assay, Simultaneously analyze multiple Th17 cytokine and chemokine biomarkers with the Th17 Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, ≥95.0% (HPLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder