Merck
  • Localized SCF and IGF-1 secretion enhances erythropoiesis in the spleen of murine embryos.
Localized SCF and IGF-1 secretion enhances erythropoiesis in the spleen of murine embryos.
Biology open (2015-04-19)
Keai Sinn Tan, Tomoko Inoue, Kasem Kulkeaw, Yuka Tanaka, Mei I Lai, Daisuke Sugiyama
ABSTRACT
Fetal spleen is a major hematopoietic site prior to initiation of bone marrow hematopoiesis. Morphologic analysis suggested erythropoietic activity in fetal spleen, but it remained unclear how erythropoiesis was regulated. To address this question, we performed flow cytometric analysis and observed that the number of spleen erythroid cells increased 18.6-fold from 16.5 to 19.5 days post-coitum (dpc). Among erythropoietic cytokines, SCF and IGF-1 were primarily expressed in hematopoietic, endothelial and mesenchymal-like fetal spleen cells. Cultures treated with SCF and/or IGF-1R inhibitors showed significantly decreased CD45-c-Kit-CD71+/-Ter119+ erythroid cells and downregulated Gata1, Klf1 and β-major globin expression. Administration of these inhibitors to pregnant mice significantly decreased the number of CD45-c-Kit-CD71+/-Ter119+ cells and downregulated β-major globin gene expression in embryos derived from these mice. We conclude that fetal spleen is a major erythropoietic site where endothelial and mesenchymal-like cells primarily accelerate erythropoietic activity through SCF and IGF-1 secretion.
Sigma-Aldrich
Eosin Y disodium salt, certified by the BSC
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Sigma-Aldrich
ISCK03, ≥98% (HPLC), solid
Sigma-Aldrich
Picropodophyllotoxin, ≥96% (HPLC), powder
Sigma-Aldrich
Eosin Y disodium salt, Dye content ≥85 %
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Hematoxylin