Merck
  • The Eukaryotic-Like Ser/Thr Kinase PrkC Regulates the Essential WalRK Two-Component System in Bacillus subtilis.
The Eukaryotic-Like Ser/Thr Kinase PrkC Regulates the Essential WalRK Two-Component System in Bacillus subtilis.
PLoS genetics (2015-06-24)
Elizabeth A Libby, Lindsie A Goss, Jonathan Dworkin
ABSTRACT
Most bacteria contain both eukaryotic-like Ser/Thr kinases (eSTKs) and eukaryotic-like Ser/Thr phosphatases (eSTPs). Their role in bacterial physiology is not currently well understood in large part because the conditions where the eSTKs are active are generally not known. However, all sequenced Gram-positive bacteria have a highly conserved eSTK with extracellular PASTA repeats that bind cell wall derived muropeptides. Here, we report that in the Gram-positive bacterium Bacillus subtilis, the PASTA-containing eSTK PrkC and its cognate eSTP PrpC converge with the essential WalRK two-component system to regulate WalR regulon genes involved in cell wall metabolism. By continuously monitoring gene expression throughout growth, we consistently find a large PrkC-dependent effect on expression of several different WalR regulon genes in early stationary phase, including both those that are activated by WalR (yocH) as well as those that are repressed (iseA, pdaC). We demonstrate that PrkC phosphorylates WalR in vitro and in vivo on a single Thr residue located in the receiver domain. Although the phosphorylated region of the receiver domain is highly conserved among several B. subtilis response regulators, PrkC displays specificity for WalR in vitro. Consistently, strains expressing a nonphosphorylatable WalR point mutant strongly reduce both PrkC dependent activation and repression of yocH, iseA, and pdaC. This suggests a model where the eSTK PrkC regulates the essential WalRK two-component signaling system by direct phosphorylation of WalR Thr101, resulting in the regulation of WalR regulon genes involved in cell wall metabolism in stationary phase. As both the eSTK PrkC and the essential WalRK two-component system are highly conserved in Gram-positive bacteria, these results may be applicable to further understanding the role of eSTKs in Gram-positive physiology and cell wall metabolism.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Monoclonal Anti-FLAG-Peroxidase antibody produced in rat, 2-4 mg/mL, clone 6F7, purified immunoglobulin
Sigma-Aldrich
Magnesium chloride solution, BioUltra, ~0.025 M in H2O
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Acrylamide, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Magnesium chloride solution, 1.00 M±0.01 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis