Merck
  • Fiber-optic immunosensor for detection of Crimean-Congo hemorrhagic fever IgG antibodies in patients.
Fiber-optic immunosensor for detection of Crimean-Congo hemorrhagic fever IgG antibodies in patients.
Analytical chemistry (2015-07-08)
Fairoz Algaar, Evgeni Eltzov, Marina M Vdovenko, Ivan Yu Sakharov, Luka Fajs, Manfred Weidmann, Ali Mirazimi, Robert S Marks
ABSTRACT
Crimean-Congo hemorrhagic fever (CCHF) is a severe viral disease with high fatality rate. CCHF virus is endemic in parts of Africa, Asia, the Middle East, and southeastern Europe. Rapid diagnostics of CCHF is vital for appropriate clinical management and prevention of secondary spread from human-to-human. Currently, diagnostics relies on real-time RT-PCR and antibody or antigen detection using ELISA. These methods require trained personnel and expensive equipment and are not appropriate for point-of-care (POC) diagnostics. Furthermore, there are no POC assays available for CCHF. We developed a fiber-optic biosensor for the detection of CCHF IgG antibodies. In order to improve sensitivity, we optimized both the bioreceptor immobilization protocol and the chemiluminescence substrate formulation. The resulting protocol showed a 100-fold greater sensitivity for detection of CCHF antibodies. Finally, we evaluated the fiber-optic biosensor with two CCHF patient sera. We showed that the fiber-optic biosensor is 10-times more sensitive than colorimetric ELISA and is able to detect both patients with high and low levels of IgG antibodies. We believe that the fiber-optic biosensor is a suitable alternative to ELISA as it is much more sensitive and makes it possible to detect a small amount of antibodies at an early stage of infection and can be integrated as a point-of-care diagnostic system of CCHF.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
SAFC
Glycine
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for ELISA, peroxidase substrate
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Methanol
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium cyanoborohydride, reagent grade, 95%
Sigma-Aldrich
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH
Sigma-Aldrich
Sodium cyanoborohydride solution, 1.0 M in THF
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
TWEEN® 20, BioXtra, viscous liquid
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, BioReagent, 36.5-38.0%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Glycine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%