Merck
  • UCP2-induced fatty acid synthase promotes NLRP3 inflammasome activation during sepsis.
UCP2-induced fatty acid synthase promotes NLRP3 inflammasome activation during sepsis.
The Journal of clinical investigation (2015-01-13)
Jong-Seok Moon, Seonmin Lee, Mi-Ae Park, Ilias I Siempos, Maria Haslip, Patty J Lee, Mijin Yun, Chun K Kim, Judie Howrylak, Stefan W Ryter, Kiichi Nakahira, Augustine M K Choi
ABSTRACT
Cellular lipid metabolism has been linked to immune responses; however, the precise mechanisms by which de novo fatty acid synthesis can regulate inflammatory responses remain unclear. The NLRP3 inflammasome serves as a platform for caspase-1-dependent maturation and secretion of proinflammatory cytokines. Here, we demonstrated that the mitochondrial uncoupling protein-2 (UCP2) regulates NLRP3-mediated caspase-1 activation through the stimulation of lipid synthesis in macrophages. UCP2-deficient mice displayed improved survival in a mouse model of polymicrobial sepsis. Moreover, UCP2 expression was increased in human sepsis. Consistently, UCP2-deficient mice displayed impaired lipid synthesis and decreased production of IL-1β and IL-18 in response to LPS challenge. In macrophages, UCP2 deficiency suppressed NLRP3-mediated caspase-1 activation and NLRP3 expression associated with inhibition of lipid synthesis. In UCP2-deficient macrophages, inhibition of lipid synthesis resulted from the downregulation of fatty acid synthase (FASN), a key regulator of fatty acid synthesis. FASN inhibition by shRNA and treatment with the chemical inhibitors C75 and cerulenin suppressed NLRP3-mediated caspase-1 activation and inhibited NLRP3 and pro-IL-1β gene expression in macrophages. In conclusion, our results suggest that UCP2 regulates the NLRP3 inflammasome by inducing the lipid synthesis pathway in macrophages. These results identify UCP2 as a potential therapeutic target in inflammatory diseases such as sepsis.
Product Description
SAFC
BIS-TRIS
SAFC
BIS-TRIS
Sigma-Aldrich
Anti-Uncoupling Protein-2 (UCP-2) (144-157) Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by trichloroacetic acid extraction
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Alanine, European Pharmacopoeia (EP) Reference Standard
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Oil Red O solution, 0.5% in isopropanol
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Poly(deoxyadenylic-thymidylic) acid sodium salt, double-stranded alternating copolymer
Sigma-Aldrich
Cerulenin, ≥98% (HPLC), from Cephalosporium caerulens
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)