Merck
  • Ikaros imposes a barrier to CD8+ T cell differentiation by restricting autocrine IL-2 production.
Ikaros imposes a barrier to CD8+ T cell differentiation by restricting autocrine IL-2 production.
Journal of immunology (Baltimore, Md. : 1950) (2014-04-30)
Shaun O'Brien, Rajan M Thomas, Gerald B Wertheim, Fuqin Zhang, Hao Shen, Andrew D Wells
ABSTRACT
Naive CD4(+) T cells require signals from the TCR and CD28 to produce IL-2, expand, and differentiate. However, these same signals are not sufficient to induce autocrine IL-2 production by naive CD8(+) T cells, which require cytokines provided by other cell types to drive their differentiation. The basis for failed autocrine IL-2 production by activated CD8(+) cells is unclear. We find that Ikaros, a transcriptional repressor that silences IL-2 in anergic CD4(+) T cells, also restricts autocrine IL-2 production by CD8(+) T cells. We find that CD8(+) T cell activation in vitro in the absence of exogenous cytokines and CD4 help leads to marked induction of Ikaros, a known repressor of the Il2 gene. Naive murine CD8 T cells haplo-insufficient for Ikzf1 failed to upregulate Ikaros, produced autocrine IL-2, and differentiated in an IL-2-dependent manner into IFN-γ-producing CTLs in response to TCR/CD28 stimulation alone. Furthermore, Ikzf1 haplo-insufficient CD8(+) T cells were more effective at controlling Listeria infection and B16 melanoma growth in vivo, and they could provide help to neighboring, non-IL-2-producing cells to differentiate into IFN-γ-producing effectors. Therefore, by repressing autocrine IL-2 production, Ikaros ensures that naive CD8(+) T cells remain dependent on licensing by APCs and CD4(+) T cells, and it may therefore act as a cell-intrinsic safeguard against inappropriate CTL differentiation and immunopathology.
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, ≥95.0% (HPLC)