Merck
  • Intestinal deletion of leptin signaling alters activity of nutrient transporters and delayed the onset of obesity in mice.
Intestinal deletion of leptin signaling alters activity of nutrient transporters and delayed the onset of obesity in mice.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2014-06-15)
Annabelle Tavernier, Jean-Baptiste Cavin, Maude Le Gall, Robert Ducroc, Raphaël G P Denis, Françoise Cluzeaud, Sandra Guilmeau, Yassine Sakar, Laurence Barbot, Nathalie Kapel, Johanne Le Beyec, Francisca Joly, Streamson Chua, Serge Luquet, Andre Bado
ABSTRACT
The importance of B-isoform of leptin receptor (LEPR-B) signaling in the hypothalamus, pancreas, or liver has been well characterized, but in the intestine, a unique site of entry for dietary nutrition into the body, it has been relatively ignored. To address this question, we characterized a mouse model deficient for LEPR-B specifically in intestinal epithelial cells (IECs). (IEC)LEPR-B-knockout (KO) and wild-type (WT) mice were generated by Cre-Lox strategy and fed a normal or high-fat diet (HFD). The analyses of the animals involved histology and immunohistochemistry of intestinal mucosa, indirect calorimetric measurements, whole-body composition, and expression and activities of nutrient transporters. (IEC)LEPR-B-KO mice exhibited a 2-fold increase in length of jejunal villi and have normal growth on a normal diet but were less susceptible (P<0.01) to HFD-induced obesity. No differences occurred in energy intake and expenditure between (IEC)LEPR-B-WT and -KO mice, but (IEC)LEPR-B-KO mice fed an HFD showed increased excreted fats (P<0.05). Activities of the Na(+)/glucose cotransporter SGLT-1 and GLUT2 were unaffected in LEPR-B-KO jejunum, while GLUT5-mediated fructose transport and PepT1-mediated peptide transport were substantially reduced (P<0.01). These data demonstrate that intestinal LEPR-B signaling is important for the onset of diet-induced obesity. They suggest that intestinal LEPR-B could be a potential per os target for prevention against obesity.
Product Description
Supelco
Hydrogen peroxide solution, for ultratrace analysis, 30 % (w/w)
Fructose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(−)-Fructose, tested according to Ph. Eur.
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Supelco
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
Millipore
D-(−)-Fructose, ≥99.0% (HPLC), suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
USP
Fructose, United States Pharmacopeia (USP) Reference Standard
Supelco
Fructose, Pharmaceutical Secondary Standard; Certified Reference Material