Merck
  • Toward anticancer immunotherapeutics: well-defined polymer-antibody conjugates for selective dendritic cell targeting.
Toward anticancer immunotherapeutics: well-defined polymer-antibody conjugates for selective dendritic cell targeting.
Macromolecular bioscience (2014-07-01)
Kristof Tappertzhofen, Monika Bednarczyk, Kaloian Koynov, Matthias Bros, Stephan Grabbe, Rudolf Zentel
ABSTRACT
This paper describes the synthesis of semitelechelic maleimide-modified N-(2-hydroxypropyl)methacrylamid) (HPMA) based polymers of narrow dispersity that can be conjugated e.g. to anti-DEC-205 antibodies affording "star-like" topologies (one antibody decorated with several polymer chains). FCS revealed a hydrodynamic diameter of R(h)  = 7.9 nm and SEC narrow dispersity (1.45). Primary in vitro studies with bone marrow derived dendritic cells (DC) show higher cellular binding and uptake rates compared to control samples. Moreover, incubating these conjugates to primary splenocytes demonstrates a much higher affinity to the primary DCs than to any other immune cell population within the spleen. This differentiation is, thereby, much more pronounced for the star-like conjugates than for conjugates made from polymers statistically modified with anti-DEC-205.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Supelco
2-Mercaptoethanol, LiChropur, ≥99.0% (GC)
Supelco
Cadaverine, analytical standard
Sigma-Aldrich
Cadaverine, purum, ≥97.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Pentafluorophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Mercaptoethanol, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Cadaverine, 95%